2-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-3,3-dimercapto-acrylonitrile

Lieferant: Apollo Scientific

PC446074-1G
APOSPC446074-1GEA 248 EUR
APOSPC446074-1G
2-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-3,3-dimercapto-acrylonitrile
2-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-3,3-dimercapto-acrylonitrile
Formel: C₉H₄ClF₃N₂S₂
Molecular Weight: 296,72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24386164
CAS-Nummer: 1431555-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR