2-Bromo-1-(4-fluoro-3-methylphenyl)ethanone

Lieferant: Apollo Scientific
Gefahr

PC446075-1G
APOSPC446075-1GEA 207 EUR
APOSPC446075-1G
2-Bromo-1-(4-fluoro-3-methylphenyl)ethanone
2-Bromo-1-(4-fluoro-3-methylphenyl)ethanone
Formel: C₉H₈BrFO
Molecular Weight: 231.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18089176
CAS-Nummer: 63529-31-7
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR