N-(6-Chloro-4-trifluoromethyl-pyridin-2-yl)-N'-[1-(2,5-dimethoxy-phenyl)-meth-(E)-ylidene]-hydrazine

Lieferant: Apollo Scientific

PC446076-100MG
APOSPC446076-100MGEA 265 EUR
APOSPC446076-100MG
N-(6-Chloro-4-trifluoromethyl-pyridin-2-yl)-N'-[1-(2,5-dimethoxy-phenyl)-meth-(E)-ylidene]-hydrazine
N-(6-Chloro-4-trifluoromethyl-pyridin-2-yl)-N'-[1-(2,5-dimethoxy-phenyl)-meth-(E)-ylidene]-hydrazine
Formel: C₁₅H₁₃ClF₃N₃O₂
Molecular Weight: 359,731 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24386175
CAS-Nummer: 1431559-91-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR