2-(6-Chloro-4-trifluoromethyl-pyridin-2-ylsulphanyl)-N-ethyl-acetamide

Lieferant: Apollo Scientific

PC446078-100MG
APOSPC446078-100MGEA 265 EUR
APOSPC446078-100MG
2-(6-Chloro-4-trifluoromethyl-pyridin-2-ylsulphanyl)-N-ethyl-acetamide
2-(6-Chloro-4-trifluoromethyl-pyridin-2-ylsulphanyl)-N-ethyl-acetamide
Formel: C₁₀H₁₀ClF₃N₂OS
Molecular Weight: 298,712 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24386177
CAS-Nummer: 1431555-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR