N'-[2-(6-Chloro-4-trifluoromethyl-pyridin-2-ylsulphanyl)-acetyl]-hydrazinecarboxylic acid ethyl ester
Lieferant: Apollo Scientific
PC446079-100MG
APOSPC446079-100MGEA
321
EUR
APOSPC446079-100MG
N'-[2-(6-Chloro-4-trifluoromethyl-pyridin-2-ylsulphanyl)-acetyl]-hydrazinecarboxylic acid ethyl ester
N'-[2-(6-Chloro-4-trifluoromethyl-pyridin-2-ylsulphanyl)-acetyl]-hydrazinecarboxylic acid ethyl ester
Formel:
C₁₁H₁₁ClF₃N₃O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD24386178 CAS-Nummer: 1431555-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...