N-Benzyl-2-(6-morpholin-4-yl-4-trifluoromethyl-pyridin-2-ylsulphanyl)-acetamide

Lieferant: Apollo Scientific

PC446080-100MG
APOSPC446080-100MGEA 328 EUR
APOSPC446080-100MG
N-Benzyl-2-(6-morpholin-4-yl-4-trifluoromethyl-pyridin-2-ylsulphanyl)-acetamide
N-Benzyl-2-(6-morpholin-4-yl-4-trifluoromethyl-pyridin-2-ylsulphanyl)-acetamide
Formel: C₁₉H₂₀F₃N₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD24386179
CAS-Nummer: 1431555-23-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR