4-(5-Nitro-6-pyrrolidin-1-yl-4-trifluoromethyl-pyridin-2-yl)-morpholine
Lieferant: Apollo Scientific
PC446081-100MG
APOSPC446081-100MGEA
265
EUR
APOSPC446081-100MG
4-(5-Nitro-6-pyrrolidin-1-yl-4-trifluoromethyl-pyridin-2-yl)-morpholine
4-(5-Nitro-6-pyrrolidin-1-yl-4-trifluoromethyl-pyridin-2-yl)-morpholine
Formel:
C₁₄H₁₇F₃N₄O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD24386180 CAS-Nummer: 1431555-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...