4-(6-Chloro-4-trifluoromethyl-pyridin-2-yloxy)-phenylamine

Lieferant: Apollo Scientific

PC446082-100MG
APOSPC446082-100MGEA 265 EUR
APOSPC446082-100MG
4-(6-Chloro-4-trifluoromethyl-pyridin-2-yloxy)-phenylamine
4-(6-Chloro-4-trifluoromethyl-pyridin-2-yloxy)-phenylamine
Formel: C₁₂H₈ClF₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD24386181
CAS-Nummer: 86575-23-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR