2-Bromo-3-(trifluoromethyl)benzoyl chloride
Lieferant: Apollo Scientific
PC446083-1G
APOSPC446083-1GEA
308
EUR
APOSPC446083-1G
2-Bromo-3-(trifluoromethyl)benzoyl chloride
2-Bromo-3-(trifluoromethyl)benzoyl chloride
Formel:
C₈H₃BrClF₃O Molecular Weight: 287,47 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185388 CAS-Nummer: 1214376-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...