5-Fluorobenzo[b]thiophene-3-carboxaldehyde 95+%

Lieferant: Apollo Scientific

PC446106-1G
APOSPC446106-1GEA 623 EUR
APOSPC446106-1G
5-Fluorobenzo[b]thiophene-3-carboxaldehyde 95+%
5-Fluorobenzo[b]thiophene-3-carboxaldehyde
Formel: C₉H₅FOS
Molecular Weight: 180.199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17168659
CAS-Nummer: 40740-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR