6-Fluorobenzo[b]thiophene-2-carboxaldehyde 95+%

Lieferant: Apollo Scientific

PC446113-1G
APOSPC446113-1GEA 0 EUR
APOSPC446113-1G
6-Fluorobenzo[b]thiophene-2-carboxaldehyde 95+%
6-Fluorobenzo[b]thiophene-2-carboxaldehyde
Formel: C₉H₅FOS
Molecular Weight: 180,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18451485
CAS-Nummer: 212078-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR