7-(Trifluoromethyl)-1H-indole-2-carboxylic acid 95+%

Lieferant: Apollo Scientific

PC446119-1G
APOSPC446119-1GEA 387 EUR
APOSPC446119-1G
7-(Trifluoromethyl)-1H-indole-2-carboxylic acid 95+%
7-(Trifluoromethyl)-1H-indole-2-carboxylic acid
Formel: C₁₀H₆F₃NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD02664424
CAS-Nummer: 172216-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR