2-Methyl-5-[4-(trifluoromethyl)phenyl]furan-3-carboxamide 95+%

Lieferant: Apollo Scientific

PC446120-1G
APOSPC446120-1GEA 497 EUR
APOSPC446120-1G
2-Methyl-5-[4-(trifluoromethyl)phenyl]furan-3-carboxamide 95+%
2-Methyl-5-[4-(trifluoromethyl)phenyl]furan-3-carboxamide
Formel: C₁₃H₁₀F₃NO₂
Molecular Weight: 269,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27937104
CAS-Nummer: 1858241-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR