4-Fluor-2-nitrobenzotrifluorid 90+%
Lieferant: Apollo Scientific
Synonyme:
4-Fluoro-2-nitro-1-(trifluoromethyl)benzene
PC446128-1G
APOSPC446128-1GEA
243
EUR
APOSPC446128-1G
4-Fluor-2-nitrobenzotrifluorid 90+%
4-Fluor-2-nitrobenzotrifluorid
Formel:
C₇H₃F₄NO₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
182289-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...