6-Methyl-4-(trifluoromethyl)pyridine-2-carbonitrile 95+%

Lieferant: Apollo Scientific

PC446131-1G
APOSPC446131-1GEA 751 EUR
APOSPC446131-1G
6-Methyl-4-(trifluoromethyl)pyridine-2-carbonitrile 95+%
6-Methyl-4-(trifluoromethyl)pyridine-2-carbonitrile
Formel: C₈H₅F₃N₂
Molecular Weight: 186,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18259812
CAS-Nummer: 451459-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR