3-Bromo-2-fluorobenzenesulphonyl chloride 95+%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-2-fluorobenzene-1-sulphonyl chloride

PC446132-1G
APOSPC446132-1GEA 142 EUR
APOSPC446132-1G
3-Bromo-2-fluorobenzenesulphonyl chloride 95+%
3-Bromo-2-fluorobenzenesulphonyl chloride
Formel: C₆H₃BrClFO₂S
Molecular Weight: 273,507 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185362
CAS-Nummer: 1214372-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR