2-Bromo-5-fluorobenzenesulphonyl chloride 95+%

Lieferant: Apollo Scientific

PC446133-1G PC446133-5G
APOSPC446133-1GEA 92.8 EUR
APOSPC446133-1G APOSPC446133-5G
2-Bromo-5-fluorobenzenesulphonyl chloride 95+%
2-Bromo-5-fluorobenzenesulphonyl chloride
Formel: C₆H₃BrClFO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD13185361
CAS-Nummer: 771-67-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR