5-Bromo-3-fluoro-2-methoxybenzaldehyde 95+%

Lieferant: Apollo Scientific

PC446134-1G
APOSPC446134-1GEA 623 EUR
APOSPC446134-1G
5-Bromo-3-fluoro-2-methoxybenzaldehyde 95+%
5-Bromo-3-fluoro-2-methoxybenzaldehyde
Formel: C₈H₆BrFO₂
Molecular Weight: 233,034 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22035163
CAS-Nummer: 470668-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR