3-(4-Fluoro-phenoxy)phenylamine 95+%

Lieferant: Apollo Scientific

PC446135-1G
APOSPC446135-1GEA 411 EUR
APOSPC446135-1G
3-(4-Fluoro-phenoxy)phenylamine 95+%
3-(4-Fluoro-phenoxy)phenylamine
Formel: C₁₂H₁₀FNO
Molecular Weight: 203.212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236585
CAS-Nummer: 203302-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR