6-Fluoropyridine-2-carbonyl chloride 95+%

Lieferant: Apollo Scientific

PC446136-1G
APOSPC446136-1GEA 369 EUR
APOSPC446136-1G
6-Fluoropyridine-2-carbonyl chloride 95+%
6-Fluoropyridine-2-carbonyl chloride
Formel: C₆H₃ClFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD04038711
CAS-Nummer: 64197-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR