2-Fluoroisonicotinic acid tert-butyl ester 95+%
Lieferant: Apollo Scientific
PC446138-1G
APOSPC446138-1GEA
598
EUR
APOSPC446138-1G
2-Fluoroisonicotinic acid tert-butyl ester 95+%
2-Fluoroisonicotinic acid tert-butyl ester
Formel:
C₁₀H₁₂FNO₂ Molecular Weight: 197,206 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18897596 CAS-Nummer: 1339544-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...