3-Fluoroisonicotinic acid tert-butyl ester 95+%

Lieferant: Apollo Scientific

PC446139-1G
APOSPC446139-1GEA 825 EUR
APOSPC446139-1G
3-Fluoroisonicotinic acid tert-butyl ester 95+%
3-Fluoroisonicotinic acid tert-butyl ester
Formel: C₁₀H₁₂FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD21114998
CAS-Nummer: 1254475-68-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR