2-Fluoronicotinic acid tert-butyl ester 95+%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 2-fluoronicotinate
PC446141-1G
APOSPC446141-1GEA
497
EUR
APOSPC446141-1G
2-Fluoronicotinic acid tert-butyl ester 95+%
2-Fluoronicotinic acid tert-butyl ester
Formel:
C₁₀H₁₂FNO₂ Molecular Weight: 197,206 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29918664 CAS-Nummer: 1501904-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...