5-Fluoropyridine-2-carboxylic acid tert-butyl ester 95+%

Lieferant: Apollo Scientific

PC446142-1G
APOSPC446142-1GEA 623 EUR
APOSPC446142-1G
5-Fluoropyridine-2-carboxylic acid tert-butyl ester 95+%
5-Fluoropyridine-2-carboxylic acid tert-butyl ester
Formel: C₁₀H₁₂FNO₂
Molecular Weight: 197,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18910562
CAS-Nummer: 1379037-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR