(E)-3-(2-Fluoro-4-hydroxyphenyl)acrylic acid 95+%

Lieferant: Apollo Scientific

PC446145-1G
APOSPC446145-1GEA 623 EUR
APOSPC446145-1G
(E)-3-(2-Fluoro-4-hydroxyphenyl)acrylic acid 95+%
(E)-3-(2-Fluoro-4-hydroxyphenyl)acrylic acid
Formel: C₉H₇FO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06797933
CAS-Nummer: 696589-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR