2-Amino-4-iodo-5-(trifluoromethyl)pyridine 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Iodo-5-(trifluoromethyl)pyridin-2-amine, 4-Iodo-5-(trifluoromethyl)pyridin-2-ylamine

PC446150-1G
APOSPC446150-1GEA 513 EUR
APOSPC446150-1G
2-Amino-4-iodo-5-(trifluoromethyl)pyridine 95+%
2-Amino-4-iodo-5-(trifluoromethyl)pyridine
Formel: C₆H₄F₃IN₂
Molecular Weight: 288,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16606194
CAS-Nummer: 1227515-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR