3-(4-Fluoro-phenoxy)propionitrile 95+%

Lieferant: Apollo Scientific

PC446153-5G
APOSPC446153-5GEA 130 EUR
APOSPC446153-5G
3-(4-Fluoro-phenoxy)propionitrile 95+%
3-(4-Fluoro-phenoxy)propionitrile
Formel: C₉H₈FNO
Molecular Weight: 165,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01569391
CAS-Nummer: 85169-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR