3-Chloro-5-fluorobenzene-1-sulphonyl chloride 95+%

Lieferant: Apollo Scientific
Gefahr

PC446155-1G
APOSPC446155-1GEA 243 EUR
APOSPC446155-1G
3-Chloro-5-fluorobenzene-1-sulphonyl chloride 95+%
3-Chloro-5-fluorobenzene-1-sulphonyl chloride
Formel: C₆H₃Cl₂FO₂S
Molecular Weight: 229,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185437
CAS-Nummer: 1131397-73-3
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR