2-(4-Fluorphenyl)pyridin 95+%

Lieferant: Apollo Scientific

PC446161-1G
APOSPC446161-1GEA 115 EUR
APOSPC446161-1G
2-(4-Fluorphenyl)pyridin 95+%
2-(4-Fluorophenyl)pyridine
Formel: C₁₁H₈FN
Lagertemperatur: Raumtemperatur
MDL: MFCD06201382
CAS-Nummer: 58861-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR