5-Fluoro-2-phenylpyridine 95+%

Lieferant: Apollo Scientific

PC446162-1G
APOSPC446162-1GEA 217 EUR
APOSPC446162-1G
5-Fluoro-2-phenylpyridine 95+%
5-Fluoro-2-phenylpyridine
Formel: C₁₁H₈FN
Molecular Weight: 173.186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044346
CAS-Nummer: 512171-81-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR