2-(2-Chloro-4-fluorophenyl)-3-oxo-butyronitrile 95+%

Lieferant: Apollo Scientific

PC446163-1G
APOSPC446163-1GEA 623 EUR
APOSPC446163-1G
2-(2-Chloro-4-fluorophenyl)-3-oxo-butyronitrile 95+%
2-(2-Chloro-4-fluorophenyl)-3-oxo-butyronitrile
Formel: C₁₀H₇ClFNO
Molecular Weight: 211,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17282112
CAS-Nummer: 1358582-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR