5-(4-Fluorophenyl)furan-2-carboxaldehyde 95+%
Lieferant: Apollo Scientific
PC446170-1G
APOSPC446170-1GEA
306
EUR
APOSPC446170-1G
5-(4-Fluorophenyl)furan-2-carboxaldehyde 95+%
5-(4-Fluorophenyl)furan-2-carboxaldehyde
Formel:
C₁₁H₇FO₂ Molecular Weight: 190,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00463034 CAS-Nummer: 33342-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...