7-Fluoro-quinazolin-2-ylamine 95+%

Lieferant: Apollo Scientific

PC446181-1G
APOSPC446181-1GEA 497 EUR
APOSPC446181-1G
7-Fluoro-quinazolin-2-ylamine 95+%
7-Fluoro-quinazolin-2-ylamine
Formel: C₈H₆FN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09264074
CAS-Nummer: 190274-01-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR