2-Chloro-4-(trifluoromethyl)pyridine 1-oxide 95+%

Lieferant: Apollo Scientific

PC446182-1G
APOSPC446182-1GEA 446 EUR
APOSPC446182-1G
2-Chloro-4-(trifluoromethyl)pyridine 1-oxide 95+%
2-Chloro-4-(trifluoromethyl)pyridine 1-oxide
Formel: C₆H₃ClF₃NO
Molecular Weight: 197.542 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28401555
CAS-Nummer: 164464-58-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR