4-Brom-2,6-difluorbenzamid 95+%

Lieferant: Apollo Scientific

PC446191-1G
APOSPC446191-1GEA 302 EUR
APOSPC446191-1G
4-Brom-2,6-difluorbenzamid 95+%
4-Brom-2,6-difluorbenzamid
Formel: C₇H₄BrF₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD13689151
CAS-Nummer: 840481-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR