4-Trifluoromethyl-L-phenylalanine 95%

Lieferant: Apollo Scientific

PC4463-10G PC4463-1G PC4463-5G
APOSPC4463-10GEA 800 EUR
APOSPC4463-10G APOSPC4463-1G APOSPC4463-5G
4-Trifluoromethyl-L-phenylalanine 95%
4-Trifluoromethyl-L-phenylalanine
Formel: C₁₀H₁₀F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00044946
CAS-Nummer: 114926-38-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR