Potassium thiophene-3-trifluoroborate
Lieferant: Apollo Scientific
Synonyme:
Potassium 3-thiophenetrifluoroborate, Potassium trifluoro(thiophen-3-yl)borate
PC4467-25G
APOSPC4467-25GEA
217
EUR
APOSPC4467-25G
Potassium thiophene-3-trifluoroborate
Potassium thiophene-3-trifluoroborate
Formel:
C₄H₃BF₃KS Molecular Weight: 190.032 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03701626 CAS-Nummer: 192863-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...