2-{[4-(Trifluoromethyl)pyridin-3-yl]thio}acetic acid 95+%

Lieferant: Apollo Scientific
Achtung

PC4479-1G
APOSPC4479-1GEA 256 EUR
APOSPC4479-1G
2-{[4-(Trifluoromethyl)pyridin-3-yl]thio}acetic acid 95+%
2-{[4-(Trifluoromethyl)pyridin-3-yl]thio}acetic acid
Formel: C₈H₆F₃NO₂S
Schmelzpunkt: 140…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319255
CAS-Nummer: 387350-44-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR