1-Butyl-2,3-dimethylimidazoliumtetrafluoroborat
Lieferant: Apollo Scientific
|
Gefahr
|
PC448000-25G
APOSPC448000-25GEA
0
EUR
APOSPC448000-25G
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborat
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate
Formel:
C₉H₁₇BF₄N₂ Schmelzpunkt: 34 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03427618 CAS-Nummer: 402846-78-0 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...