1-Butyl-3-(trifluoromethyl)pyrazole

Lieferant: Apollo Scientific

PC448002-1G
APOSPC448002-1GEA 0 EUR
APOSPC448002-1G
1-Butyl-3-(trifluoromethyl)pyrazole
1-Butyl-3-(trifluoromethyl)pyrazole
Formel: C₈H₁₁F₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD21668277
CAS-Nummer: 1426958-36-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR