1-(3-Bromo-2-fluorophenyl)ethanol

Lieferant: Apollo Scientific

PC448003-1G
APOSPC448003-1GEA 771 EUR
APOSPC448003-1G
1-(3-Bromo-2-fluorophenyl)ethanol
1-(3-Bromo-2-fluorophenyl)ethanol
Formel: C₈H₈BrFO
Molecular Weight: 219.053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16140247
CAS-Nummer: 1221715-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR