4-Bromo-1-butyl-3-(trifluoromethyl)pyrazole

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-1-butyl-3-(trifluoromethyl)-1H-pyrazole

PC448005-1G
APOSPC448005-1GEA 85.7 EUR
APOSPC448005-1G
4-Bromo-1-butyl-3-(trifluoromethyl)pyrazole
4-Bromo-1-butyl-3-(trifluoromethyl)pyrazole
Formel: C₈H₁₀BrF₃N₂
Molecular Weight: 271,078 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24448781
CAS-Nummer: 1437794-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR