4-Bromo-1-isobutyl-3-(trifluoromethyl)pyrazole

Lieferant: Apollo Scientific

PC448006-1G
APOSPC448006-1GEA 430 EUR
APOSPC448006-1G
4-Bromo-1-isobutyl-3-(trifluoromethyl)pyrazole
4-Bromo-1-isobutyl-3-(trifluoromethyl)pyrazole
Formel: C₈H₁₀BrF₃N₂
Molecular Weight: 271,078 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24448782
CAS-Nummer: 1437794-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR