3,5-Difluor-4-formylphenylboronsäure

Lieferant: Apollo Scientific

Synonyme: (3,5-Difluoro-4-formylphenyl)boronic acid

PC448009-1G
APOSPC448009-1GEA 77 EUR
APOSPC448009-1G
3,5-Difluor-4-formylphenylboronsäure
3,5-Difluor-4-formylphenylboronsäure
Formel: C₇H₅BF₂O₃
Molecular Weight: 185,9206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784344
CAS-Nummer: 870718-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR