4-(4-Fluorophenyl)-7-hydroxy-2H-chromen-2-one

Lieferant: Apollo Scientific

Synonyme: 2H-1-Benzopyran-2-one,4-(4-fluorophenyl)-7-hydroxy-

PC448013-1G
APOSPC448013-1GEA 277 EUR
APOSPC448013-1G
4-(4-Fluorophenyl)-7-hydroxy-2H-chromen-2-one
4-(4-Fluorophenyl)-7-hydroxy-2H-chromen-2-one
Formel: C₁₅H₉FO₃
Molecular Weight: 256,229 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11112064
CAS-Nummer: 850881-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR