6-Fluoro-3-methylbenzo[d]isoxazol-5-amine hydrochloride

Lieferant: Apollo Scientific

PC448017-1G
APOSPC448017-1GEA 815 EUR
APOSPC448017-1G
6-Fluoro-3-methylbenzo[d]isoxazol-5-amine hydrochloride
6-Fluoro-3-methylbenzo[d]isoxazol-5-amine hydrochloride
Formel: C₈H₇FN₂O·ClH
Molecular Weight: 202,613 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16036053
CAS-Nummer: 1243313-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR