2-Fluoro-3-hydroxybenzaldehyde 98%

Lieferant: Apollo Scientific

PC448018-5G
APOSPC448018-5GEA 156 EUR
APOSPC448018-5G
2-Fluoro-3-hydroxybenzaldehyde 98%
2-Fluoro-3-hydroxybenzaldehyde
Formel: C₇H₅FO₂
Molecular Weight: 140.112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11110191
CAS-Nummer: 103438-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR