1-Hexyl-3-methylimidazoliumtetrafluoroborat
Lieferant: Apollo Scientific
Synonyme:
1-n-Hexyl-3-methylimidazolium tetrafluoroborate, [HMIM] [BF4]
PC448020-100G
APOSPC448020-100GEA
154
EUR
APOSPC448020-100G
1-Hexyl-3-methylimidazoliumtetrafluoroborat
1-Hexyl-3-methylimidazolium tetrafluoroborate
Formel:
C₁₀H₁₉BF₄N₂ Molecular Weight: 254,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095459 CAS-Nummer: 244193-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...