1-Isopropyl-3-(trifluoromethyl)pyrazole-5-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-Isopropyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)boronic acid

PC448022-1G
APOSPC448022-1GEA 109 EUR
APOSPC448022-1G
1-Isopropyl-3-(trifluoromethyl)pyrazole-5-boronic acid
1-Isopropyl-3-(trifluoromethyl)pyrazole-5-boronic acid
Formel: C₇H₁₀BF₃N₂O₂
Molecular Weight: 221,973 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD24448764
CAS-Nummer: 1361380-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR