1-Isobutyl-3-(trifluoromethyl)pyrazole

Lieferant: Apollo Scientific

PC448023-1G
APOSPC448023-1GEA 323 EUR
APOSPC448023-1G
1-Isobutyl-3-(trifluoromethyl)pyrazole
1-Isobutyl-3-(trifluoromethyl)pyrazole
Formel: C₈H₁₁F₃N₂
Molecular Weight: 192,182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21668199
CAS-Nummer: 1437795-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR